missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7125893
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3213200MG 200 mg
C32131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3213200MG Supplier TCI America Supplier No. C3213200MG

Chemical Identifiers

CAS 957061-11-9
Molecular Formula C7H5BClF3O2
Molecular Weight (g/mol) 224.37
MDL Number MFCD09258745
InChI Key NEHJDQKHVZYUAA-UHFFFAOYSA-N
Synonym 2-Chloro-3-(trifluoromethyl)benzeneboronic Acid
PubChem CID 44558143
IUPAC Name [2-chloro-3-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C(=CC=C1)C(F)(F)F)Cl)(O)O

Specifications

Color White
Quantity 200 mg
Formula Weight 224.37
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)