missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Formula Weight | 224.37 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Chloro-3-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
| TSCA | No |