missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-pyridinemethanol 98.0+%, TCI America™

Catalog No. C25905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25905G 5 g
C25901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25905G Supplier TCI America Supplier No. C25905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42330-59-6
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD01935961
InChI Key HMPDWSBKPCOQDW-UHFFFAOYSA-N
Synonym 2-chloropyridin-3-yl methanol, 2-chloro-3-pyridinyl methanol, 2-chloro-3-pyridinylmethanol, 2-chloro-3-pyridinemethanol, 2-chloro-3-hydroxymethyl pyridine, 2-chloro-pyridin-3-yl-methanol, 3-pyridinemethanol, 2-chloro, 2-chloro-3-hydroxymethylpyridine
PubChem CID 2763647
IUPAC Name (2-chloropyridin-3-yl)methanol
SMILES OCC1=CC=CN=C1Cl

Specifications

Melting Point 63°C
Boiling Point 112°C
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-3-pyridinemethanol