accessibility menu, dialog, popup

UserName

HONCl

CAS RN 42330-59-6

IUPAC Name: (2-chloropyridin-3-yl)methanol
Synonyms: (2-chloropyridin-3-yl)methanol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: HMPDWSBKPCOQDW-UHFFFAOYSA-N
SMILES: OCC1=CC=CN=C1Cl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)