missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-pyridinemethanol, 97%, Thermo Scientific Chemicals

Catalog No. AAH6334103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6334103 1 g
AAH6334106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6334103 Supplier Thermo Scientific Chemicals Supplier No. H6334103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42330-59-6
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD01935961
InChI Key HMPDWSBKPCOQDW-UHFFFAOYSA-N
Synonym 2-chloropyridin-3-yl methanol, 2-chloro-3-pyridinyl methanol, 2-chloro-3-pyridinylmethanol, 2-chloro-3-pyridinemethanol, 2-chloro-3-hydroxymethyl pyridine, 2-chloro-pyridin-3-yl-methanol, 3-pyridinemethanol, 2-chloro, 2-chloro-3-hydroxymethylpyridine
PubChem CID 2763647
IUPAC Name (2-chloropyridin-3-yl)methanol
SMILES OCC1=CC=CN=C1Cl

Specifications

Melting Point 61°C to 65°C (decomposition)
Boiling Point 112°C (0.2 mmHg)
Quantity 1 g
Beilstein 1525378
Formula Weight 143.57
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-3-pyridinemethanol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only