missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitropyridine, 99+%, Thermo Scientific™

$177.60 - $605.88

Chemical Identifiers

CAS 5470-18-8
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00006232
InChI Key UUOLETYDNTVQDY-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
PubChem CID 79613
IUPAC Name 2-chloro-3-nitropyridine
SMILES C1=CC(=C(N=C1)Cl)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC109730250
View Documents Promotion Details
N/A 25g Glass bottle
Each for $177.60
Only null left
 
AC109731000
View Documents Promotion Details
N/A 100g Glass bottle
Each for $605.88
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5470-18-8
158.54
UUOLETYDNTVQDY-UHFFFAOYSA-N
79613
C1=CC(=C(N=C1)Cl)[N+](=O)[O-]
C5H3ClN2O2
MFCD00006232
pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
2-chloro-3-nitropyridine

Specifications

5470-18-8
100.0
Authentic
C5H3ClN2O2
Cl(C5H3N)NO2
20, I, 82
Solubility in water: insoluble
C1=CC(=C(N=C1)Cl)[N+](=O)[O-]
158.54
158.54
2-chloro-3-nitropyridine
99.0
185°C
99% min. (HPLC)
1.4540 to 1.4560
MFCD00006232
pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
UUOLETYDNTVQDY-UHFFFAOYSA-N
2-chloro-3-nitropyridine
79613
99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 226-799-8

SDS
Documents

Documents

RUO – Research Use Only