missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitropyridine, 99+%

Catalog No. AC109731000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109731000 100 g Glass bottle
AC109730250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109731000 Supplier Thermo Scientific Chemicals Supplier No. 109731000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5470-18-8 | C5H3ClN2O2 | 158.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5470-18-8
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00006232
InChI Key UUOLETYDNTVQDY-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
PubChem CID 79613
IUPAC Name 2-chloro-3-nitropyridine
SMILES C1=CC(=C(N=C1)Cl)[N+](=O)[O-]

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 100.0°C to 103.0°C
Color Yellow
Density 185g/mL
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.4540 to 1.4560
Linear Formula Cl(C5H3N)NO2
Quantity 100 g
Beilstein 20, I, 82
Solubility Information Solubility in water: insoluble
Formula Weight 158.54
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-3-nitropyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 226-799-8

RUO – Research Use Only