missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3-methylbenzoic Acid 98.0+%, TCI America™
$85.75 - $255.38
Chemical Identifiers
| CAS | 15068-35-6 |
|---|---|
| Molecular Formula | C8H7ClO2 |
| Molecular Weight (g/mol) | 170.59 |
| MDL Number | MFCD00079736 |
| InChI Key | LWOKLXMNGXXORN-UHFFFAOYSA-N |
| Synonym | 2-Chloro-m-toluic Acid |
| PubChem CID | 139879 |
| IUPAC Name | 2-chloro-3-methylbenzoic acid |
| SMILES | CC1=CC=CC(C(O)=O)=C1Cl |
Chemical Identifiers
| 15068-35-6 | |
| 170.59 | |
| LWOKLXMNGXXORN-UHFFFAOYSA-N | |
| 139879 | |
| CC1=CC=CC(C(O)=O)=C1Cl |
| C8H7ClO2 | |
| MFCD00079736 | |
| 2-Chloro-m-toluic Acid | |
| 2-chloro-3-methylbenzoic acid |
Specifications
| 15068-35-6 | |
| C8H7ClO2 | |
| 5 g | |
| LWOKLXMNGXXORN-UHFFFAOYSA-N | |
| 2-chloro-3-methylbenzoic acid | |
| 139879 | |
| ≥98.0% (GC,T) | |
| 2-Chloro-3-methylbenzoic Acid |
| 143°C | |
| MFCD00079736 | |
| 2-Chloro-m-toluic Acid | |
| CC1=CC=CC(C(O)=O)=C1Cl | |
| 170.59 | |
| 170.59 | |
| Crystalline Powder |
Safety and Handling
TSCA : No