missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3-methoxypropionic Acid 98.0+%, TCI America™
$335.52 - $1284.33
Chemical Identifiers
| CAS | 2544-05-0 |
|---|---|
| Molecular Formula | C4H7ClO3 |
| Molecular Weight (g/mol) | 138.547 |
| MDL Number | MFCD03093624 |
| InChI Key | SSWGTERYCJZJPQ-UHFFFAOYSA-N |
| PubChem CID | 14269106 |
| IUPAC Name | 2-chloro-3-methoxypropanoic acid |
| SMILES | COCC(C(=O)O)Cl |
Chemical Identifiers
| 2544-05-0 | |
| 138.547 | |
| SSWGTERYCJZJPQ-UHFFFAOYSA-N | |
| 2-chloro-3-methoxypropanoic acid |
| C4H7ClO3 | |
| MFCD03093624 | |
| 14269106 | |
| COCC(C(=O)O)Cl |
Specifications
| 2544-05-0 | |
| 155°C | |
| MFCD03093624 | |
| 3265 | |
| COCC(C(=O)O)Cl | |
| 138.547 | |
| 138.55 | |
| Liquid |
| Yellow | |
| C4H7ClO3 | |
| 5 g | |
| SSWGTERYCJZJPQ-UHFFFAOYSA-N | |
| 2-chloro-3-methoxypropanoic acid | |
| 14269106 | |
| ≥98.0% (GC,T) | |
| 2-Chloro-3-methoxypropionic Acid |
Safety and Handling
TSCA : No