missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-hexylthiophene 98.0+%, TCI America™

Catalog No. c26135g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26135G Supplier TCI America Supplier No. C26135G
Only null left

Chemical Identifiers

CAS 817181-75-2
Molecular Formula C10H15ClS
Molecular Weight (g/mol) 202.74
MDL Number MFCD18413738
InChI Key BSWSVVYUCAKNDK-UHFFFAOYSA-N
PubChem CID 56973699
IUPAC Name 2-chloro-3-hexylthiophene
SMILES CCCCCCC1=C(SC=C1)Cl

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
Formula Weight 202.74
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-hexylthiophene
TSCA No
Recommended Storage Refrigerator