missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluorobenzoic Acid 97.0+%, TCI America™

Catalog No. C21281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21281G 1 g
C21285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21281G Supplier TCI America Supplier No. C21281G
Only null left

Chemical Identifiers

CAS 102940-86-3
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00973903
InChI Key WJYAYXKXZNITAZ-UHFFFAOYSA-N
Synonym 2-chloro-3-fluorobenzoicacid, 2-chloro-3-fluoro-benzoic acid, benzoic acid, 2-chloro-3-fluoro, pubchem1369, ksc493s0f, buttpark 19\01-64, 2-chloranyl-3-fluoranyl-benzoic acid
PubChem CID 302620
IUPAC Name 2-chloro-3-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)F)Cl)C(=O)O

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 1 g
Formula Weight 174.56
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-fluorobenzoic Acid