Learn More
2-Chloro-3,4-dimethoxypyridine, 97%, Thermo Scientific Chemicals
$296.58 - $296.58
Chemical Identifiers
| CAS | 101664-59-9 |
|---|---|
| Molecular Formula | C7H9Cl2NO2 |
| Molecular Weight (g/mol) | 210.054 |
| MDL Number | MFCD03265283 |
| InChI Key | AOBQXXVCWDAJFE-UHFFFAOYSA-N |
| Synonym | 2-chloro-3,4-dimethoxypyridine hydrochloride, 2-chloro-3,4-dimethoxypyridine hcl, 2-chloro-3,4-dimethoxypyridine-hydrogen chloride 1/1 |
| PubChem CID | 24728279 |
| IUPAC Name | 2-chloro-3,4-dimethoxypyridine;hydrochloride |
| SMILES | COC1=C(C(=NC=C1)Cl)OC.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3369703
|
Thermo Scientific Chemicals
H3369703 |
1 g |
Each for $296.58
|
|
|||||
Description
2-Chloro-3,4-dimethoxypyridine is used in the preparation of 3, 4-dimethoxy-pyridine by using nickel(II) chloride hexahydrate and triphenylphosphine as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-3,4-dimethoxypyridine is used in the preparation of 3, 4-dimethoxy-pyridine by using nickel(II) chloride hexahydrate and triphenylphosphine as a catalyst.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Chemical Identifiers
| 101664-59-9 | |
| 210.054 | |
| AOBQXXVCWDAJFE-UHFFFAOYSA-N | |
| 24728279 | |
| COC1=C(C(=NC=C1)Cl)OC.Cl |
| C7H9Cl2NO2 | |
| MFCD03265283 | |
| 2-chloro-3,4-dimethoxypyridine hydrochloride, 2-chloro-3,4-dimethoxypyridine hcl, 2-chloro-3,4-dimethoxypyridine-hydrogen chloride 1/1 | |
| 2-chloro-3,4-dimethoxypyridine;hydrochloride |
Specifications
| 101664-59-9 | |
| C7H9Cl2NO2 | |
| 1 g | |
| AOBQXXVCWDAJFE-UHFFFAOYSA-N | |
| 2-chloro-3,4-dimethoxypyridine;hydrochloride | |
| 24728279 | |
| 97% |
| 37°C to 42°C | |
| MFCD03265283 | |
| 2-chloro-3,4-dimethoxypyridine hydrochloride, 2-chloro-3,4-dimethoxypyridine hcl, 2-chloro-3,4-dimethoxypyridine-hydrogen chloride 1/1 | |
| COC1=C(C(=NC=C1)Cl)OC.Cl | |
| 210.054 | |
| 173.6 | |
| 2-Chloro-3,4-dimethoxypyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only