CAS RN 101664-59-9
IUPAC Name:
2-chloro-3,4-dimethoxypyridine hydrochloride
Synonyms:
2-chloro-3,4-dimethoxypyridine hydrochloride
Molecular Weight (g/mol):
210.05
Molecular Formula:
C7H9Cl2NO2
InChi Key:
AOBQXXVCWDAJFE-UHFFFAOYSA-N
SMILES:
Cl.COC1=C(OC)C(Cl)=NC=C1
1
–
1
of
1
results
1
Promotions Available
2-Chloro-3,4-dimethoxypyridine, 97%, Thermo Scientific Chemicals
2-Chloro-3,4-dimethoxypyridine, 97%, Thermo Scientific Chemicals
CAS: 101664-59-9 Molecular Formula: C7H9Cl2NO2 Molecular Weight (g/mol): 210.054 MDL Number: MFCD03265283 InChI Key: AOBQXXVCWDAJFE-UHFFFAOYSA-N Synonym: 2-chloro-3,4-dimethoxypyridine hydrochloride,2-chloro-3,4-dimethoxypyridine hcl,2-chloro-3,4-dimethoxypyridine-hydrogen chloride 1/1 PubChem CID: 24728279 IUPAC Name: 2-chloro-3,4-dimethoxypyridine;hydrochloride SMILES: COC1=C(C(=NC=C1)Cl)OC.Cl