missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3',4'-dihydroxyacetophenone, 97%
Description
2-Chloro-3',4'-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.
Solubility
Soluble in DMSO, methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.
Solubility
Soluble in DMSO, methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 172°C to 176°C |
| Quantity | 500 g |
| Beilstein | 2092660 |
| Solubility Information | Soluble in DMSO,methanol. |
| Formula Weight | 186.59 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Chloro-3',4'-dihydroxyacetophenone |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 202-754-8 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only