missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3',4'-dihydroxyacetophenone, 97%

Catalog No. AAB2349614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2349614 25 g
AAB2349622 100 g
AAB2349636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2349614 Supplier Thermo Scientific Chemicals Supplier No. B2349614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-40-1 | C8H7ClO3 | 186.591 g/mol

2-Chloro-3',4'-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

Solubility
Soluble in DMSO, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 99-40-1
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00002200
InChI Key LWTJEJCZJFZKEL-UHFFFAOYSA-N
Synonym 2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone
PubChem CID 66834
ChEBI CHEBI:51844
IUPAC Name 2-chloro-1-(3,4-dihydroxyphenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CCl)O)O

Specifications

Melting Point 172°C to 176°C
Quantity 25 g
Beilstein 2092660
Solubility Information Soluble in DMSO,methanol.
Formula Weight 186.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-3',4'-dihydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-754-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only