missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3',4'-difluoroacetophenone 98.0+%, TCI America™

Catalog No. C32155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32155G 5 g
C32151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32155G Supplier TCI America Supplier No. C32155G

Chemical Identifiers

CAS 51336-95-9
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
MDL Number MFCD03966888
InChI Key VMEDAWUIKFAFJQ-UHFFFAOYSA-N
PubChem CID 2392321
IUPAC Name 2-chloro-1-(3,4-difluorophenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CCl)F)F

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 190.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-3′,4′-difluoroacetophenone