accessibility menu, dialog, popup

UserName

CAS RN 51336-95-9

IUPAC Name: 2-chloro-1-(3,4-difluorophenyl)ethan-1-one
Synonyms: 2-chloro-1-(3,4-difluorophenyl)ethanone
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: VMEDAWUIKFAFJQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1F)C(=O)CCl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results