missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylbutane 97.0+%, TCI America™

Catalog No. C020025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C020025ML 25 mL
C0200500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C020025ML Supplier TCI America Supplier No. C020025ML

Chemical Identifiers

CAS 594-36-5
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00000818
InChI Key CRNIHJHMEQZAAS-UHFFFAOYSA-N
Synonym tert-amyl chloride, butane, 2-chloro-2-methyl, 2-methyl-2-chlorobutane, tert-pentyl chloride, 1,1-dimethylpropyl chloride, tertiary pentyl chloride, t-pentyl chloride, acmc-209mdc, 2-chloro-2-methyl butane, 2-chloro-2-methyl-butane
PubChem CID 61143
IUPAC Name 2-chloro-2-methylbutane
SMILES CCC(C)(C)Cl

Specifications

Color Colorless
Boiling Point 86°C
Quantity 25 mL
UN Number 1107
Formula Weight 106.59
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylbutane