accessibility menu, dialog, popup

UserName

H3CCH3CH3Cl

CAS RN 594-36-5

IUPAC Name: 2-chloro-2-methylbutane
Synonyms: tert-amyl chloride
Molecular Weight (g/mol): 106.59
Molecular Formula: C5H11Cl
InChi Key: CRNIHJHMEQZAAS-UHFFFAOYSA-N
SMILES: CCC(C)(C)Cl
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)