missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-deoxy-D-glucose, MilliporeSigma™ Supelco™

Catalog No. 111013146
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013146 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013146 Supplier Merck Emd Millipore Supplier No. C20310MG
Featured Industries: Food and Beverages

Specifications

Packaging Glass Bottle
CAS 14685-79-1
Physical Form Solid
Linear Formula C6H11 ClO5
MDL Number MFCD01317996
Melting Point 139°C to 145°C
Synonym 2-CIDG
Type Analytical Standard
Quantity 10 mg
Formula Weight 198.60
Grade Analytical Standard
Show More Show Less