missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2',6'-dimethylacetanilide, 99%

Catalog No. AAL0272709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0272709 10 g
AAL0272718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0272709 Supplier Thermo Scientific Chemicals Supplier No. L0272709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1131-01-7 | C10H12ClNO | 197.662 g/mol

2-Chloro-2',6'-dimethylacetanilide is used in the determination of the enantiomers of tocainide in blood plasma by gas-liquid chromatography with electron-capture detection. It is a nonsteroidal antinflammatory drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in Chloroform, Methanol. Insoluble in water.

Notes
Keep the container tightly closed in a cool, dry and well ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1131-01-7
Molecular Formula C10H12ClNO
Molecular Weight (g/mol) 197.662
MDL Number MFCD00000926
InChI Key FPQQSNUTBWFFLB-UHFFFAOYSA-N
Synonym 2-chloro-n-2,6-dimethylphenyl acetamide, 2-chloro-2',6'-dimethylacetanilide, n-chloroacetyl-2,6-dimethylaniline, 2-chloro-2',6'-acetoxylidide, chloroaceto-2,6-xylidide, acetamide, 2-chloro-n-2,6-dimethylphenyl, chloroacetamido-2,6-xylidine, 1-chloroacetylamino-2,6-dimethylbenzene, n-2,6-dimethylphenyl-2-chloroacetamide, unii-5d2b69oapx
PubChem CID 70798
IUPAC Name 2-chloro-N-(2,6-dimethylphenyl)acetamide
SMILES CC1=C(C(=CC=C1)C)NC(=O)CCl

Specifications

Melting Point 143°C to 148°C
Quantity 10 g
Beilstein 781309
Solubility Information Soluble in Chloroform,Methanol. Insoluble in water.
Formula Weight 197.67
Percent Purity 99%
Chemical Name or Material 2-Chloro-2',6'-dimethylacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-460-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only