missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene, 98%

Catalog No. aah6422103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6422103 1 g
AAH6422106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6422103 Supplier Thermo Scientific Chemicals Supplier No. H6422103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 654-55-7 | C8H2ClF6NO2 | 293.549 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 654-55-7
Molecular Formula C8H2ClF6NO2
Molecular Weight (g/mol) 293.549
MDL Number MFCD04972677
InChI Key OCRFDZOBCZLPRB-UHFFFAOYSA-N
Synonym 2-chloro-1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl-2-chloronitrobenzene, 3,5-bis trifluoromethyl-2-chloro-nitrobenzene, 2-chloro-3,5-di trifluoromethyl nitrobenzene, 2-chloro-3,5-bis trifluoromethyl nitrobenzene, pubchem16471, 2,4-bis-trifluoromethyl-6-nitrochlorobenzene, 2-chloro-3,5-bis-trifluoromethyl-nitrobenzene, 3,5-bis-trifluoromethyl-2-chloro-nitrobenzene, 2-chloro-1-nitro-3,5-bis-trifluoromethyl-benzene
PubChem CID 10803688
IUPAC Name 2-chloro-1-nitro-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C(=C1C(F)(F)F)Cl)[N+](=O)[O-])C(F)(F)F

Specifications

Quantity 1 g
Beilstein 2888862
Formula Weight 293.55
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only