missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-methylpyridinium Iodide 98.0+%, TCI America™

Catalog No. C090325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C090325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C090325G Supplier TCI America Supplier No. C090325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14338-32-0
Molecular Formula C6H7ClIN
Molecular Weight (g/mol) 255.48
MDL Number MFCD00011984
InChI Key ABFPKTQEQNICFT-UHFFFAOYSA-M
Synonym 2-chloro-1-methylpyridinium iodide, 2-chloro-1-methylpyridin-1-ium iodide, pyridinium, 2-chloro-1-methyl-, iodide, unii-rlq88nyz68, rlq88nyz68, 2-chloro-1-methylpyridiniumiodide, 2-chloro-1-methyl-pyridinium iodide, pubchem16659, acmc-1brmd, ksc494o1l
PubChem CID 167069
IUPAC Name 2-chloro-1-methylpyridin-1-ium iodide
SMILES [I-].C[N+]1=CC=CC=C1Cl

Specifications

Quantity 25 g
Formula Weight 255.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1-methylpyridinium Iodide