accessibility menu, dialog, popup

UserName

CAS RN 14338-32-0

IUPAC Name: 2-chloro-1-methylpyridin-1-ium iodide
Synonyms: 2-chloro-1-methylpyridin-1-ium iodide
Molecular Weight (g/mol): 255.48
Molecular Formula: C6H7ClIN
InChi Key: ABFPKTQEQNICFT-UHFFFAOYSA-M
SMILES: [I-].C[N+]1=CC=CC=C1Cl
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)