Learn More
2-Chloro-1-methylpyridinium iodide, 97%
CAS: 14338-32-0 | C6H7ClIN | 255.48 g/mol
Supplier: Thermo Scientific Chemicals A1282014
Description
2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.
Solubility
Soluble in Methanol (50 mg/ml). Insoluble in water.
Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
| 2-Chloro-1-methylpyridinium iodide | |
| ∼200°C (decomposition) | |
| MFCD00011984 | |
| 3572320 | |
| 14,6301 | |
| Soluble in Methanol (50mg/ml). Insoluble in water. | |
| [I-].C[N+]1=CC=CC=C1Cl | |
| 255.48 | |
| 255.49 |
| 14338-32-0 | |
| C6H7ClIN | |
| 25 g | |
| Light and moisture sensitive | |
| 2-chloro-1-methylpyridinium iodide, 2-chloro-1-methylpyridin-1-ium iodide, pyridinium, 2-chloro-1-methyl-, iodide, unii-rlq88nyz68, rlq88nyz68, 2-chloro-1-methylpyridiniumiodide, 2-chloro-1-methyl-pyridinium iodide, pubchem16659, acmc-1brmd, ksc494o1l | |
| ABFPKTQEQNICFT-UHFFFAOYSA-M | |
| 2-chloro-1-methylpyridin-1-ium;iodide | |
| 167069 | |
| 97% |
Chemical Identifiers
| 14338-32-0 | |
| 255.48 | |
| ABFPKTQEQNICFT-UHFFFAOYSA-M | |
| 167069 | |
| [I-].C[N+]1=CC=CC=C1Cl |
| C6H7ClIN | |
| MFCD00011984 | |
| 2-chloro-1-methylpyridinium iodide, 2-chloro-1-methylpyridin-1-ium iodide, pyridinium, 2-chloro-1-methyl-, iodide, unii-rlq88nyz68, rlq88nyz68, 2-chloro-1-methylpyridiniumiodide, 2-chloro-1-methyl-pyridinium iodide, pubchem16659, acmc-1brmd, ksc494o1l | |
| 2-chloro-1-methylpyridin-1-ium;iodide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-288-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only