missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-methylbenzimidazole, Thermo Scientific Chemicals

Catalog No. p-7055072
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6355903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6355903 Supplier Thermo Scientific Chemicals Supplier No. H6355903

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1849-02-1
Molecular Formula C8H7ClN2
Molecular Weight (g/mol) 166.61
MDL Number MFCD00769821
InChI Key UXZYKSFMGDWHGJ-UHFFFAOYSA-N
Synonym 2-chloro-1-methyl-1h-benzo d imidazole, 2-chloro-1-methyl-1h-benzimidazole, 2-chloro-1-methyl-1h-benzoimidazole, 2-chloro-1-methyl-1,3-benzodiazole, 1-methyl-1h-benzo d imidazol-2-yl chloride, 2-chloro-1-methyl-1h-1,3-benzodiazole, 1-methyl-2-chlorobenzimidazole, 2-chloro-1-methyl-benzimidazole, 1h-benzimidazole, 2-chloro-1-methyl, acmc-209em7
PubChem CID 663080
IUPAC Name 2-chloro-1-methylbenzimidazole
SMILES CN1C(Cl)=NC2=CC=CC=C12

Specifications

Color White to Yellow
Quantity 1 g
Formula Weight 166.61
Physical Form Solid
Chemical Name or Material 2-Chloro-1-methylbenzimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only