accessibility menu, dialog, popup

UserName

CAS RN 1849-02-1

IUPAC Name: 2-chloro-1-methyl-1H-1,3-benzodiazole
Synonyms: 2-chloro-1-methyl-1,3-benzodiazole
Molecular Weight (g/mol): 166.61
Molecular Formula: C8H7ClN2
InChi Key: UXZYKSFMGDWHGJ-UHFFFAOYSA-N
SMILES: CN1C(Cl)=NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results