Learn More
2-Chloro-1-iodo-4-nitrobenzene, 98%
CAS: 41252-96-4 | C6H3ClINO2 | 283.449 g/mol
Supplier: Thermo Scientific Chemicals A1934006
Description
3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
| 2-Chloro-1-iodo-4-nitrobenzene | |
| 97°C to 101°C | |
| MFCD00024589 | |
| 2502411 | |
| 3-chloro-4-iodonitrobenzene, 3-chloro-4-iodo-nitrobenzene, 3-chloro-4-iodo-nitro-benzene, benzene, 2-chloro-1-iodo-4-nitro, 2-chlor-1-iod-4-nitrobenzol, acmc-20a36k, 3-chloro-4-iodonitro benzene, 1-iodo-2-chloro-4-nitrobenzene, 2-chloro-1-iodo-4-nitro-benzene, 1-chloro-2-iodo-5-nitrobenzene | |
| FQXFHRSYMORXKN-UHFFFAOYSA-N | |
| 2-chloro-1-iodo-4-nitrobenzene | |
| 96640 | |
| 98% |
| 41252-96-4 | |
| C6H3ClINO2 | |
| 5 g | |
| Light sensitive | |
| Slightly soluble in water. | |
| C1=CC(=C(C=C1[N+](=O)[O-])Cl)I | |
| 283.449 | |
| 283.45 |
Chemical Identifiers
| 41252-96-4 | |
| 283.449 | |
| FQXFHRSYMORXKN-UHFFFAOYSA-N | |
| 96640 | |
| C1=CC(=C(C=C1[N+](=O)[O-])Cl)I |
| C6H3ClINO2 | |
| MFCD00024589 | |
| 3-chloro-4-iodonitrobenzene, 3-chloro-4-iodo-nitrobenzene, 3-chloro-4-iodo-nitro-benzene, benzene, 2-chloro-1-iodo-4-nitro, 2-chlor-1-iod-4-nitrobenzol, acmc-20a36k, 3-chloro-4-iodonitro benzene, 1-iodo-2-chloro-4-nitrobenzene, 2-chloro-1-iodo-4-nitro-benzene, 1-chloro-2-iodo-5-nitrobenzene | |
| 2-chloro-1-iodo-4-nitrobenzene |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 255-278-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only