missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-iodo-4-nitrobenzene, 98%

Catalog No. AAA1934003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1934003 1 g
AAA1934006 5 g
AAA1934014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1934003 Supplier Thermo Scientific Chemicals Supplier No. A1934003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41252-96-4 | C6H3ClINO2 | 283.449 g/mol

3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Chemical Identifiers

CAS 41252-96-4
Molecular Formula C6H3ClINO2
Molecular Weight (g/mol) 283.449
MDL Number MFCD00024589
InChI Key FQXFHRSYMORXKN-UHFFFAOYSA-N
Synonym 3-chloro-4-iodonitrobenzene, 3-chloro-4-iodo-nitrobenzene, 3-chloro-4-iodo-nitro-benzene, benzene, 2-chloro-1-iodo-4-nitro, 2-chlor-1-iod-4-nitrobenzol, acmc-20a36k, 3-chloro-4-iodonitro benzene, 1-iodo-2-chloro-4-nitrobenzene, 2-chloro-1-iodo-4-nitro-benzene, 1-chloro-2-iodo-5-nitrobenzene
PubChem CID 96640
IUPAC Name 2-chloro-1-iodo-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)I

Specifications

Melting Point 97°C to 101°C
Quantity 1 g
Beilstein 2502411
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 283.45
Percent Purity 98%
Chemical Name or Material 2-Chloro-1-iodo-4-nitrobenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 255-278-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only