missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-(ethoxymethyl)imidazole, 97%, Thermo Scientific™

Catalog No. AAH5187003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5187003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5187003 Supplier Thermo Scientific Chemicals Supplier No. H5187003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-1-(ethoxymethyl)imidazole is used as a pharmaceutical intermediate. It is also used in the synthesis of a new class of antifungal agents. Further, it is employed in Suzuki-Miyaura coupling reactions.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 850429-55-9
Molecular Formula C6H9ClN2O
Molecular Weight (g/mol) 160.60
MDL Number MFCD03788569
InChI Key DONYAQVHLUVENE-UHFFFAOYSA-N
Synonym 2-chloro-1-ethoxymethyl-1h-imidazole, 2-chloro-1-ethoxymethyl imidazole, 1h-imidazole,2-chloro-1-ethoxymethyl, pubchem9009, acmc-209pzq, 2-chloranyl-1-ethoxymethyl imidazole
PubChem CID 2773326
IUPAC Name 2-chloro-1-(ethoxymethyl)imidazole
SMILES CCOCN1C=CN=C1Cl

Specifications

Quantity 1 g
Formula Weight 160.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-1-(ethoxymethyl)imidazole
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures