CAS RN 850429-55-9
IUPAC Name:
2-chloro-1-(ethoxymethyl)-1H-imidazole
Synonyms:
2-chloro-1-(ethoxymethyl)imidazole
Molecular Weight (g/mol):
160.6
Molecular Formula:
C6H9ClN2O
InChi Key:
DONYAQVHLUVENE-UHFFFAOYSA-N
SMILES:
CCOCN1C=CN=C1Cl
1
–
1
of
1
results
1
Promotions Available
2-Chloro-1-(ethoxymethyl)imidazole, 97%, Thermo Scientific™
2-Chloro-1-(ethoxymethyl)imidazole, 97%, Thermo Scientific™
CAS: 850429-55-9 Molecular Formula: C6H9ClN2O Molecular Weight (g/mol): 160.60 MDL Number: MFCD03788569 InChI Key: DONYAQVHLUVENE-UHFFFAOYSA-N Synonym: 2-chloro-1-ethoxymethyl-1h-imidazole,2-chloro-1-ethoxymethyl imidazole,1h-imidazole,2-chloro-1-ethoxymethyl,pubchem9009,acmc-209pzq,2-chloranyl-1-ethoxymethyl imidazole PubChem CID: 2773326 IUPAC Name: 2-chloro-1-(ethoxymethyl)imidazole SMILES: CCOCN1C=CN=C1Cl