missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,4-benzoquinone, 95%

Catalog No. p-84909
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC331220050 5 g Glass bottle
AC331220250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC331220050 Supplier Thermo Scientific Chemicals Supplier No. 331220050
Only null left

CAS: 695-99-8 | C6H3ClO2 | 142.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 695-99-8
Molecular Formula C6H3ClO2
Molecular Weight (g/mol) 142.54
MDL Number MFCD00075260
InChI Key WOGWYSWDBYCVDY-UHFFFAOYSA-N
Synonym 2-chloro-1,4-benzoquinone, chloroquinone, monochloroquinone, 2-chloroquinone, chloro-p-benzoquinone, 2-chloro-p-benzoquinone, monochloro-p-benzoquinone, chloro-1,4-benzoquinone, p-benzoquinone, 2-chloro, 2,5-cyclohexadiene-1,4-dione, 2-chloro
PubChem CID 69671
IUPAC Name 2-chlorocyclohexa-2,5-diene-1,4-dione
SMILES ClC1=CC(=O)C=CC1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 52.0°C to 57.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Formula Weight 142.54
Percent Purity 95%
Physical Form Crystals
Chemical Name or Material 2-Chloro-1, 4-benzoquinone, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only