missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dimethylimidazolinium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7125959
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C34441G 1 g
C34445G 5 g
2 options

Catalog No. C34441G

Supplier: TCI America C34441G

Only null left

Chemical Identifiers

CAS 153433-26-2
Molecular Formula C5H10BClF4N2
Molecular Weight (g/mol) 220.40
MDL Number MFCD09039291
InChI Key UPLXKEIRISBKRM-UHFFFAOYSA-N
IUPAC Name 2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN1CC[N+](C)=C1Cl

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 1 g
Formula Weight 220.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1,3-dimethylimidazolinium Tetrafluoroborate
TSCA No
Recommended Storage Refrigerator