missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dimethylimidazolinium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. p-7125105
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16515G 5 g
C165125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16515G Supplier TCI America Supplier No. C16515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101385-69-7
Molecular Formula C5H10ClF6N2P
Molecular Weight (g/mol) 278.563
MDL Number MFCD00191914
InChI Key CNAKHAGVVMOXFE-UHFFFAOYSA-N
Synonym 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium hexafluorophosphate v, 2-chloro-1,3-dimethylimidazolinium hexafluorophosphate, 2-chloro-1,3-dimethylimidazolidium hexafluorophosphate, 2-chloro-1,3-dimethylimidazolidinium hexafluorophosphate, cip, 2-chloro-1,3-dimethylimidazolidinium hexafluorophosphate, 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium hexafluorophosphate, 2-chloro-4,5-dihydro-1,3-dimethyl-1h-imidazolium hexafluorophosphate, 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium; hexafluoro-lambda5-phosphanuide, 2-chloro-1,3-dimethylimidazolidinium hexafluorophosphate, purum t
PubChem CID 16211675
IUPAC Name 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium;hexafluorophosphate
SMILES CN1CC[N+](=C1Cl)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 233°C
Color White
Quantity 5 g
Formula Weight 278.56
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1,3-dimethylimidazolinium Hexafluorophosphate