accessibility menu, dialog, popup

UserName

CAS RN 101385-69-7

IUPAC Name: 2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium hexafluorophosphate
Molecular Weight (g/mol): 278.56
Molecular Formula: C5H10ClF6N2P
InChi Key: CNAKHAGVVMOXFE-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CN1CC[N+](C)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results