missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3,2-dioxaphospholane 97.0+%, TCI America™

Catalog No. p-7124858
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12155G 5 g
C121525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12155G Supplier TCI America Supplier No. C12155G
Only null left

Chemical Identifiers

CAS 822-39-9
Molecular Formula C2H4ClO2P
Molecular Weight (g/mol) 126.476
MDL Number MFCD00014511
InChI Key OLSFRDLMFAOSIA-UHFFFAOYSA-N
PubChem CID 69973
IUPAC Name 2-chloro-1,3,2-dioxaphospholane
SMILES C1COP(O1)Cl

Specifications

Melting Point -28°C
Color Colorless
Boiling Point 47°C
Quantity 5 g
UN Number 3265
Synonym ethylene chlorophosphite, 1,3,2-dioxaphospholane, 2-chloro, ethylene cyclo-chlorophosphite, acmc-209pnx, 1,3,2-dioxaphospholane,2-chloro, 2-chloro-1,3,2-dioxaphospholane, ethylene phosphorochloridite 2-chloro-1,3,2-dioxap, inchi=1/c2h4clo2p/c3-6-4-1-2-5-6/h1-2h
Formula Weight 126.48
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-1,3,2-dioxaphospholane
EINECSNumber (5)-3573
TSCA Yes
Recommended Storage Refrigerator