missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Carboxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$91.33 - $91.33
Chemical Identifiers
| CAS | 149105-19-1 |
|---|---|
| Molecular Formula | C7H7BO4 |
| MDL Number | MFCD01318118 |
| InChI Key | KWNPRVWFJOSGMZ-UHFFFAOYSA-N |
| PubChem CID | 2733985 |
Specifications
| 149105-19-1 | |
| White-Yellow | |
| MFCD01318118 | |
| 2-carboxyphenylboronic acid, 2-dihydroxyboranyl benzoic acid, 2-carboxybenzeneboronic acid, 2-dihydroxyboronyl benzoic acid, 2-dihydroxyboryl benzoic acid, carboxyphenylboronic acid, benzoic acid, 2-borono, o-carboxyphenylboronic acid, 2-carboxyphenylboronicacid, borobenzoesaure | |
| B(C1=CC=CC=C1C(=O)O)(O)O | |
| 165.939 | |
| 165.94 | |
| 2-Carboxyphenylboronic Acid (contains varying amounts of Anhydride) |
| 162°C | |
| C7H7BO4 | |
| 1 g | |
| KWNPRVWFJOSGMZ-UHFFFAOYSA-N | |
| 2-boronobenzoic acid | |
| 2733985 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator