missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. c25011g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25011G Supplier TCI America Supplier No. C25011G
Only null left

Chemical Identifiers

CAS 149105-19-1
Molecular Formula C7H7BO4
MDL Number MFCD01318118
InChI Key KWNPRVWFJOSGMZ-UHFFFAOYSA-N
PubChem CID 2733985

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 1 g
Synonym 2-carboxyphenylboronic acid, 2-dihydroxyboranyl benzoic acid, 2-carboxybenzeneboronic acid, 2-dihydroxyboronyl benzoic acid, 2-dihydroxyboryl benzoic acid, carboxyphenylboronic acid, benzoic acid, 2-borono, o-carboxyphenylboronic acid, 2-carboxyphenylboronicacid, borobenzoesaure
SMILES B(C1=CC=CC=C1C(=O)O)(O)O
IUPAC Name 2-boronobenzoic acid
Molecular Weight (g/mol) 165.939
Formula Weight 165.94
Physical Form Crystalline Powder
Chemical Name or Material 2-Carboxyphenylboronic Acid (contains varying amounts of Anhydride)
TSCA No
Recommended Storage Refrigerator