missing translation for 'onlineSavingsMsg'
Learn More

(2-Carboxyethyl)triphenylphosphonium chloride, 98%

Catalog No. AAB2560906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2560906 5 g
AAB2560914 25 g
AAB2560922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2560906 Supplier Thermo Scientific Chemicals Supplier No. B2560906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36626-29-6 | C21H20ClO2P | 370.81 g/mol

2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 36626-29-6
Molecular Formula C21H20ClO2P
Molecular Weight (g/mol) 370.81
MDL Number MFCD00185663
InChI Key GALLWJZTZYJVSL-UHFFFAOYSA-N
Synonym 2-carboxyethyl triphenylphosphonium chloride, 2-carboxyethyl tripheylphosphonium chloride, 2-carboxyethyl-triphenylphosphonium chloride, phosphonium, 2-carboxyethyl triphenyl-, chloride, 2-carboxyethyl triphenylphosphanium chloride, acmc-1agne, 2-carboxyethyl triphenyl phosphonium chloride, beta-carboxyethyltriphenylphosphonium chloride, 2-carboxyethyl triphenyl-phosphonium chloride
PubChem CID 10270534
SMILES [Cl-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 197°C to 201°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in dimethyl sulfoxide and methanol.
IUPAC Name 2-carboxyethyl(triphenyl)phosphanium;chloride
Formula Weight 370.81
Percent Purity 98%
Chemical Name or Material (2-Carboxyethyl)triphenylphosphonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only