missing translation for 'onlineSavingsMsg'
Learn More

(2-Carboxyethyl)dimethylsulfonium Bromide 98.0+%, TCI America™

Catalog No. C12415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C12415G Supplier TCI America Supplier No. C12415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20986-22-5
Molecular Formula C5H11BrO2S
Molecular Weight (g/mol) 215.11
MDL Number MFCD00142887
InChI Key UKHXPHLUQGMQOF-UHFFFAOYSA-N
PubChem CID 19881914
IUPAC Name (2-carboxyethyl)dimethylsulfanium bromide
SMILES [Br-].C[S+](C)CCC(O)=O

Specifications

Melting Point 112°C
Color White
Quantity 5 g
Formula Weight 215.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Carboxyethyl)dimethylsulfonium Bromide