accessibility menu, dialog, popup

UserName

H3CSCH3OOHBr

CAS RN 20986-22-5

IUPAC Name: (2-carboxyethyl)dimethylsulfanium bromide
Synonyms: (2-carboxyethyl)dimethylsulfanium bromide
Molecular Weight (g/mol): 215.11
Molecular Formula: C5H11BrO2S
InChi Key: UKHXPHLUQGMQOF-UHFFFAOYSA-N
SMILES: [Br-].C[S+](C)CCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results