Learn More
Chemical Identifiers
| CAS | 59337-92-7 |
|---|---|
| Molecular Formula | C6H5ClO4S2 |
| Molecular Weight (g/mol) | 240.67 |
| MDL Number | MFCD00068160 |
| InChI Key | PJVJBDAUWILEOG-UHFFFAOYSA-N |
| Synonym | methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride |
| PubChem CID | 2736733 |
| IUPAC Name | methyl 3-chlorosulfonylthiophene-2-carboxylate |
| SMILES | COC(=O)C1=C(C=CS1)S(Cl)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC398610010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 59337-92-7 | |
| 240.67 | |
| PJVJBDAUWILEOG-UHFFFAOYSA-N | |
| 2736733 | |
| COC(=O)C1=C(C=CS1)S(Cl)(=O)=O |
| C6H5ClO4S2 | |
| MFCD00068160 | |
| methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride | |
| methyl 3-chlorosulfonylthiophene-2-carboxylate |
Specifications
| 59337-92-7 | |
| 100.0 | |
| 94% min. (HPLC) | |
| MFCD00068160 | |
| PJVJBDAUWILEOG-UHFFFAOYSA-N | |
| methyl 3-chlorosulfonylthiophene-2-carboxylate | |
| 2736733 | |
| 95% |
| 94.0 | |
| Authentic | |
| C6H5ClO4S2 | |
| methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride | |
| COC(=O)C1=C(C=CS1)S(Cl)(=O)=O | |
| 240.67 | |
| 240.69 | |
| 2-Carbomethoxy-3-thiophenesulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only