missing translation for 'onlineSavingsMsg'
Learn More

2-Carbomethoxy-3-thiophenesulfonyl chloride, 95%, Thermo Scientific™

Catalog No. AC398610010
Change view
Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398610010 1g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC398610010 Supplier Thermo Scientific Chemicals Supplier No. 398610010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59337-92-7
Molecular Formula C6H5ClO4S2
Molecular Weight (g/mol) 240.67
MDL Number MFCD00068160
InChI Key PJVJBDAUWILEOG-UHFFFAOYSA-N
Synonym methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride
PubChem CID 2736733
IUPAC Name methyl 3-chlorosulfonylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)S(Cl)(=O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1g
Formula Weight 240.69
Percent Purity 95%
Chemical Name or Material 2-Carbomethoxy-3-thiophenesulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only