missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Carbobenzoxyamino)-1-ethanol 97.0+%, TCI America™
$119.36 - $469.74
Chemical Identifiers
| CAS | 77987-49-6 |
|---|---|
| Molecular Formula | C10H13NO3 |
| Molecular Weight (g/mol) | 195.22 |
| MDL Number | MFCD00191060 |
| InChI Key | SAGINAGERRNGGV-UHFFFAOYSA-N |
| Synonym | benzyl n-2-hydroxyethyl carbamate, z-glycinol, benzyl 2-hydroxyethyl carbamate, n-z-ethanolamine, 2-cbz-amino-1-ethanol, benzyl 2-hydroxyethylcarbamate, 2-z-amino ethanol, n-2-hydroxyethyl phenylmethoxy carboxamide, n-2-hydroxyethyl carbamic acid benzyl ester |
| PubChem CID | 280458 |
| IUPAC Name | benzyl N-(2-hydroxyethyl)carbamate |
| SMILES | OCCNC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 77987-49-6 | |
| 195.22 | |
| SAGINAGERRNGGV-UHFFFAOYSA-N | |
| 280458 | |
| OCCNC(=O)OCC1=CC=CC=C1 |
| C10H13NO3 | |
| MFCD00191060 | |
| benzyl n-2-hydroxyethyl carbamate, z-glycinol, benzyl 2-hydroxyethyl carbamate, n-z-ethanolamine, 2-cbz-amino-1-ethanol, benzyl 2-hydroxyethylcarbamate, 2-z-amino ethanol, n-2-hydroxyethyl phenylmethoxy carboxamide, n-2-hydroxyethyl carbamic acid benzyl ester | |
| benzyl N-(2-hydroxyethyl)carbamate |
Specifications
| 60°C | |
| 215°C | |
| C10H13NO3 | |
| benzyl n-2-hydroxyethyl carbamate, z-glycinol, benzyl 2-hydroxyethyl carbamate, n-z-ethanolamine, 2-cbz-amino-1-ethanol, benzyl 2-hydroxyethylcarbamate, 2-z-amino ethanol, n-2-hydroxyethyl phenylmethoxy carboxamide, n-2-hydroxyethyl carbamic acid benzyl ester | |
| OCCNC(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 280458 | |
| ≥97.0% (GC) | |
| 2-(Carbobenzoxyamino)-1-ethanol |
| 77987-49-6 | |
| White | |
| MFCD00191060 | |
| SAGINAGERRNGGV-UHFFFAOYSA-N | |
| benzyl N-(2-hydroxyethyl)carbamate | |
| 195.22 | |
| 195.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No