missing translation for 'onlineSavingsMsg'
Learn More

2-Carbamoylbenzeneboronic acid, 96%

Catalog No. p-7034940
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52642MD 250 mg
AAH5264203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52642MD Supplier Thermo Scientific Chemicals Supplier No. H52642MD
Only null left

CAS: 380430-54-6 | C7H8BNO3 | 164.955 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 380430-54-6
Molecular Formula C7H8BNO3
Molecular Weight (g/mol) 164.955
MDL Number MFCD02179454
InChI Key LBWJTKOVBMVJJX-UHFFFAOYSA-N
Synonym 2-aminocarbonylphenylboronic acid, 2-carbamoylphenyl boronic acid, 2-carbamoylbenzeneboronic acid, 2-aminocarbonyl benzeneboronic acid, 2-boronobenzamide, boronic acid, 2-aminocarbonyl phenyl, 2-aminocarbonylphenyl boronic acid, 2-aminocarbonyl phenylboronic acid, boronic acid,b-2-aminocarbonyl phenyl, pubchem1766
PubChem CID 2737809
IUPAC Name (2-carbamoylphenyl)boronic acid
SMILES B(C1=CC=CC=C1C(=O)N)(O)O

Specifications

Melting Point 226°C to 230°C
Quantity 250 mg
Formula Weight 164.96
Percent Purity 96%
Chemical Name or Material 2-Carbamoylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only