accessibility menu, dialog, popup

UserName

CAS RN 380430-54-6

IUPAC Name: (2-carbamoylphenyl)boronic acid
Synonyms: 2-carbamoylphenylboronic acid
Molecular Weight (g/mol): 164.96
Molecular Formula: C7H8BNO3
InChi Key: LBWJTKOVBMVJJX-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results