missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butylfuran 98.0+%, TCI America™
$128.62 - $395.17
Chemical Identifiers
| CAS | 4466-24-4 |
|---|---|
| Molecular Formula | C8H12O |
| Molecular Weight (g/mol) | 124.183 |
| MDL Number | MFCD00047071 |
| InChI Key | NWZIYQNUCXUJJJ-UHFFFAOYSA-N |
| Synonym | furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane |
| PubChem CID | 20534 |
| IUPAC Name | 2-butylfuran |
| SMILES | CCCCC1=CC=CO1 |
Chemical Identifiers
| 4466-24-4 | |
| 124.183 | |
| NWZIYQNUCXUJJJ-UHFFFAOYSA-N | |
| 20534 | |
| CCCCC1=CC=CO1 |
| C8H12O | |
| MFCD00047071 | |
| furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane | |
| 2-butylfuran |
Specifications
| 4466-24-4 | |
| 59°C | |
| MFCD00047071 | |
| 1993 | |
| NWZIYQNUCXUJJJ-UHFFFAOYSA-N | |
| 2-butylfuran | |
| 20534 | |
| ≥98.0% (GC) | |
| 2-Butylfuran |
| Yellow | |
| C8H12O | |
| 5 g | |
| furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane | |
| CCCCC1=CC=CO1 | |
| 124.183 | |
| 124.18 | |
| Liquid |
Safety and Handling
TSCA : No