missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Butylamino)ethanol 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 199°C |
| Quantity | 25 mL |
| UN Number | 2735 |
| Formula Weight | 117.19 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-(Butylamino)ethanol |
Safety and Handling
| EINECSNumber | (2)-0295 |
| RTECSNumber | KK0175000 |
| TSCA | Yes |