missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran 98.0+%, TCI America™

Catalog No. B49381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49381G 1 g
B49385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49381G Supplier TCI America Supplier No. B49381G
Only null left

Chemical Identifiers

CAS 1951-26-4
Molecular Formula C19H16I2O3
Molecular Weight (g/mol) 546.143
MDL Number MFCD02675787
InChI Key PNFMEGSMKIHDFZ-UHFFFAOYSA-N
Synonym (2-Butyl-3-benzofuranyl)(4-hydroxy-3,5-diiodophenyl)methanone
PubChem CID 74769
ChEBI CHEBI:79661
IUPAC Name (2-butyl-1-benzofuran-3-yl)-(4-hydroxy-3,5-diiodophenyl)methanone
SMILES CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)O)I

Specifications

Melting Point 145°C
Quantity 1 g
Formula Weight 546.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran