CAS RN 1951-26-4
IUPAC Name:
4-(2-butyl-1-benzofuran-3-carbonyl)-2,6-diiodophenol
Synonyms:
4-(2-butyl-1-benzofuran-3-carbonyl)-2,6-diiodophenol
Molecular Weight (g/mol):
546.14
Molecular Formula:
C19H16I2O3
InChi Key:
PNFMEGSMKIHDFZ-UHFFFAOYSA-N
SMILES:
CCCCC1=C(C(=O)C2=CC(I)=C(O)C(I)=C2)C2=CC=CC=C2O1
1
–
1
of
1
results
1
2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran 98.0+%, TCI America™
CAS: 1951-26-4 Molecular Formula: C19H16I2O3 Molecular Weight (g/mol): 546.143 MDL Number: MFCD02675787 InChI Key: PNFMEGSMKIHDFZ-UHFFFAOYSA-N Synonym: (2-Butyl-3-benzofuranyl)(4-hydroxy-3,5-diiodophenyl)methanone PubChem CID: 74769 ChEBI: CHEBI:79661 IUPAC Name: (2-butyl-1-benzofuran-3-yl)-(4-hydroxy-3,5-diiodophenyl)methanone SMILES: CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)O)I